2-Isobutyl-3-methoxypyrazine 98%

Supplier: Thermo Scientific Chemicals
Warning

A13680-03 A13680-06
CAAAA13680-03EA 90.06 CAD
CAAAA13680-03 CAAAA13680-06 CAAAA13680-14
2-Isobutyl-3-methoxypyrazine 98%
2-Isobutyl-3-methoxypyrazine

Purity: 98% CAS No.: 24683-00-9 Molecular Formula: C9H14N2O Formula Weight: 166.22 Flash Point: 80°(176°F) Refractive Index: 1.49 MDL No.: MFCD00006128

Formula: C₉H₁₄N₂O
Boiling Pt: 237 °C (1013 hPa)
Melting Pt: 107 °C
Density: 0.986
Flash Pt: 80 °C(176 °F)
Storage Temperature: Ambient
MDL Number: MFCD00006128
CAS Number: 24683-00-9
EINECS: 246-402-1

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR