2-Amino-4,6-dimethylpyrimidine 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
4,6-Dimethylpyrimidin-2-ylamine, 4,6-Dimethylpyrimidin-2-amine
A13674-14
A13674-22
CAAAA13674-14EA
66.2
CAD
CAAAA13674-14
CAAAA13674-22
2-Amino-4,6-dimethylpyrimidine 98%
2-Amino-4,6-dimethylpyrimidine
Purity: 98%
CAS No.: 767-15-7
Molecular Formula: C6H9N3
Formula Weight: 123.16
Melting Point: 150-155°
Beilstein Reference: 110735
Formula:
C₆H₉N₃ MW: 123.16 g/mol Melting Pt: 150…155 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006102 CAS Number: 767-15-7 EINECS: 212-181-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...