2,4-Difluoroaniline 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A13673-14
A13673-22
A13673-36
CAAAA13673-14EA
94.48
CAD
CAAAA13673-14
CAAAA13673-22
CAAAA13673-36
2,4-Difluoroaniline 99%
2,4-Difluoroaniline
Purity: 99%
CAS No.: 367-25-9
Molecular Formula: C6H5F2N
Formula Weight: 129.11
Flash Point: 62°(143°F)
Refractive Index: 1.5070
Odor: Characteristic
Melting Point: -8 to -7°
Boiling Point: 168-170°
Beilstein Reference: 2802556
Merck Reference: 14,3147
UN No.: 2941
MDL No.: MFCD00007648
Formula:
C₆H₅F₂N Boiling Pt: 168…170 °C Melting Pt: –8 °C to –7 °C Density: 1.281 Flash Pt: 62 °C (143 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00007648 CAS Number: 367-25-9 EINECS: 206-687-5 UN: 2941 ADR: 6.1,III Merck Index: 14,3147 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...