3-Chlorophenol 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A13671-22
A13671-18
A13671-30
CAAAAA13671-0BEA
1905.02
CAD
CAAAAA13671-0B
CAAAAA13671-22
CAAAA13671-18
CAAAA13671-30
3-Chlorophenol 98+%
3-Chlorophenol
Purity: 98+%
CAS No.: 108-43-0
Molecular Formula: C6H5ClO
Formula Weight: 128.56
Flash Point: 120°(248°F)
Refractive Index: 1.5660
Melting Point: 30-34°
Boiling Point: 214°
Beilstein Reference: 1634401
Merck Reference: 14,2154
UN No.: 2020
MDL No.: MFCD00002256
Formula:
C₆H₅ClO MW: 128.56 g/mol Boiling Pt: 214 °C Melting Pt: 30…34 °C Density: 1.245 Flash Pt: 120 °C (248 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002256 CAS Number: 108-43-0 EINECS: 203-582-6 UN: 2020 ADR: 6.1,III Merck Index: 14,2154 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...