2-Nitro-p-toluidine 98+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A13666-0E
A13666-22
A13666-36
CAAAA13666-0EEA
410.8
CAD
CAAAA13666-0E
CAAAA13666-22
CAAAA13666-36
2-Nitro-p-toluidine 98+%
2-Nitro-p-toluidine
Purity: 98+%
CAS No.: 89-62-3
Molecular Formula: C7H8N2O2
Formula Weight: 152.15
Flash Point: 157°(314°F)
Odor: Characteristic
Boiling Point: 169°/21mm
Beilstein Reference: 879506
UN No.: 2660
MDL No.: MFCD00007907
Formula:
C₇H₈N₂O₂ Boiling Pt: 169 °C/21 mm Melting Pt: 115…118 °C Density: 1.31 Flash Pt: 157 °C (314 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00007907 CAS Number: 89-62-3 EINECS: 201-924-9 UN: 2660 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...