3-Fluorophthalic acid 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A13656-06
A13656-14
CAAAA13656-03EA
42.77
CAD
CAAAA13656-03
CAAAA13656-06
CAAAA13656-14
3-Fluorophthalic acid 97%
3-Fluorophthalic acid
Purity: 97%
CAS No.: 1583-67-1
Molecular Formula: C8H5FO4
Formula Weight: 184.12
Odor: Odorless
Melting Point: ca 170° dec.
Beilstein Reference: 2616684
MDL No.: MFCD00039524
Formula:
C₈H₅FO₄ Melting Pt: ∼ 170 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00039524 CAS Number: 1583-67-1 EINECS: 216-433-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...