3-(4-Methoxyphenyl)propionic acid 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
3-(4-Methoxyphenyl)propanoic acid
A13655-14
A13655-22
CAAAA13655-14EA
273.34
CAD
CAAAA13655-14
CAAAA13655-22
3-(4-Methoxyphenyl)propionic acid 98%
3-(4-Methoxyphenyl)propionic acid
Purity: 98%
CAS No.: 1929-29-9
Molecular Formula: C10H12O3
Formula Weight: 180.20
Melting Point: 102-106°
Beilstein Reference: 1953326
Formula:
C₁₀H₁₂O₃ MW: 180.2 g/mol Boiling Pt: 319 °C (1013 hPa) Melting Pt: 102…106 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002777 CAS Number: 1929-29-9 EINECS: 217-678-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...