Octanophenone 98%

Supplier: Thermo Scientific Chemicals

A13652-06 A13652-14
CAAAA13652-06EA 80.26 CAD
CAAAA13652-06 CAAAA13652-14
Octanophenone 98%
Octanophenone

Purity: 98% CAS No.: 1674-37-9 Molecular Formula: C14H20O Formula Weight: 204.31 Flash Point: 110°(230°F) Refractive Index: 1.5040 Melting Point: 22-24° Boiling Point: 285° Beilstein Reference: 1868534 MDL No.: MFCD00003554

Formula: C₆H₅C(O)(CH₂)₆CH₃
Boiling Pt: 285 °C
Melting Pt: 22…24 °C
Density: 0.938
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00003554
CAS Number: 1674-37-9
EINECS: 216-817-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7cd0f209a607b8cbbf9da6901aa068a648d8d286&vid=aaab188f5f60b7daeb700e46748ab2e0885cb4ae&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR