5-Bromoisatin 90+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A13641-18
A13641-22
A13641-36
CAAAAA13641-18EA
124.82
CAD
CAAAAA13641-18
CAAAA13641-22
CAAAA13641-36
5-Bromoisatin 90+%
5-Bromoisatin
Purity: 90+%
CAS No.: 87-48-9
Molecular Formula: C8H4BrNO2
Formula Weight: 226.03
Melting Point: ca 250° dec.
Beilstein Reference: 383760
Formula:
C₈H₄BrNO₂ Melting Pt: ∼ 250 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00149345 CAS Number: 87-48-9 EINECS: 201-747-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...