2-Bromothioanisole 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Bromophenyl methyl sulfide, 1-Bromo-2-(methylthio)benzene, 2-Bromophenyl methyl sulphide, (2-Bromophenyl)(methyl)sulfane, (2-Bromophenyl)(methyl)sulphane

A13632-03 A13632-06 A13632-14
CAAAA13632-03EA 55.3 CAD
CAAAA13632-03 CAAAA13632-06 CAAAA13632-14
2-Bromothioanisole 98%
2-Bromothioanisole

Purity: 98% CAS No.: 19614-16-5 Molecular Formula: C7H7BrS Formula Weight: 203.11 Flash Point: 110°(230°F) Refractive Index: 1.6330 Melting Point: -24° Boiling Point: 140-141°/20mm Beilstein Reference: 2243716 MDL No.: MFCD00000066

Formula: C₇H₇BrS
MW: 203.1 g/mol
Boiling Pt: 140…141 °C/20 mm
Melting Pt: –24 °C
Density: 1.524
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000066
CAS Number: 19614-16-5
EINECS: 243-183-4
UN: 3334
ADR: 9,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR