3-Hydroxybenzoic acid 99%

Supplier: Thermo Scientific Chemicals
Warning

A13628-22 A13628-36
CAAAA13628-22EA 87.53 CAD
CAAAA13628-22 CAAAA13628-36
3-Hydroxybenzoic acid 99%
3-Hydroxybenzoic acid

Purity: 99% CAS No.: 99-06-9 Molecular Formula: C7H6O3 Formula Weight: 138.12 Flash Point: 220°(428°F) Odor: Odorless Beilstein Reference: 508160 MDL No.: MFCD00002506

Formula: C₇H₆O₃
Boiling Pt: 346 °C (1013 hPa)
Melting Pt: 201…203 °C
Density: 1,473 g/cm³ (20 °C)
Flash Pt: 220 °C (428 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002506
CAS Number: 99-06-9
EINECS: 202-726-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR