Trifluoroacetic anhydride 99+%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Trifluoracetic anhydride, TFAA

A13614-14 A13614-0E A13614-22 A13614-36
CAAAAA13614-14EA 46.14 CAD
CAAAAA13614-14 CAAAA13614-0E CAAAA13614-22 CAAAA13614-36
Trifluoroacetic anhydride 99+%
Trifluoroacetic anhydride

Purity: 99+% CAS No.: 407-25-0 Molecular Formula: C4F6O3 Formula Weight: 210.03 Storage Temperature: 2°C to 8°C Refractive Index: 1.2690 Melting Point: -65° Boiling Point: 39-40° Beilstein Reference: 746197 UN No.: 3265 MDL No.: MFCD00000416

Formula: C₄F₆O₃
MW: 210.03 g/mol
Boiling Pt: 39.4
Melting Pt: -65
Density: 1.503
Storage Temperature: Refrigerator
MDL Number: MFCD00000416
CAS Number: 407-25-0
EINECS: 206-982-9
UN: 3265
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR