2-Chlorobenzoylacetonitrile 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A13611-22
A13611-06
A13611-14
CAAAAA13611-22EA
663.6
CAD
CAAAAA13611-22
CAAAA13611-06
CAAAA13611-14
2-Chlorobenzoylacetonitrile 95%
2-Chlorobenzoylacetonitrile
Purity: 98%
CAS No.: 40018-25-5
Molecular Formula: C9H6ClNO
Formula Weight: 179.61
Melting Point: 54-58°
Beilstein Reference: 2413204
Formula:
C₉H₆ClNO Melting Pt: 54…58 °C Storage Temperature: Ambient |
MDL Number:
MFCD00051624 CAS Number: 40018-25-5 UN: 3439 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...