2-Bromobenzoic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

A13608-0B A13608-18 A13608-30
CAAAA13608-0BEA 935.36 CAD
CAAAA13608-0B CAAAA13608-18 CAAAA13608-30
2-Bromobenzoic acid 98%
2-Bromobenzoic acid

Purity: 98% CAS No.: 88-65-3 Molecular Formula: C7H5BrO2 Formula Weight: 201.02 Melting Point: 146-150° Beilstein Reference: 971266

Formula: C₇H₅BrO₂
Boiling Pt: 296 °C (1013 hPa)
Melting Pt: 146…150 °C
Density: 1.930
Flash Pt: ≥ 100 °C
Storage Temperature: Ambient
MDL Number: MFCD00002402
CAS Number: 88-65-3
EINECS: 201-848-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR