Salicylonitrile 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: o-Cyanophenol, o-Hydroxybenzonitrile, 2-Cyanophenol, 2-Hydroxybenzonitrile

A13606-06 A13606-14 A13606-22
CAAAA13606-06EA 69.36 CAD
CAAAA13606-06 CAAAA13606-14 CAAAA13606-22
Salicylonitrile 98%
Salicylonitrile

Purity: 98% CAS No.: 611-20-1 Molecular Formula: C7H5NO Formula Weight: 119.12 Melting Point: 92-95° Boiling Point: 149°/14mm MDL No.: MFCD00002145

Formula: C₇H₅NO
Boiling Pt: 149 °C/14 mm
Melting Pt: 92…95 °C
Density: 1.1052 g/cm³ (99.6 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002145
CAS Number: 611-20-1
EINECS: 210-259-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR