2,4-Dimethylthiazole 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A13604-14
A13604-22
CAAAA13604-14EA
149.15
CAD
CAAAA13604-14
CAAAA13604-22
2,4-Dimethylthiazole 99%
2,4-Dimethylthiazole
MDL: MFCD00014509
Beilstein: 106414
Slightly soluble in water
Purity: 99%
CAS No.: 541-58-2
Molecular Formula: C5H7NS
Formula Weight: 113.18
Flash Point: 42°(107°F)
Refractive Index: 1.5090
Odor: Characteristic
Boiling Point: 145-147°
Solubility: Slightly soluble in water
Beilstein Reference: 106414
UN No.: 1993
MDL No.: MFCD00014509
Formula:
C₅H₇NS MW: 113.18 g/mol Boiling Pt: 145…147 °C Density: 1.057 Flash Pt: 42 °C (107 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00014509 CAS Number: 541-58-2 EINECS: 208-786-9 UN: 1993 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...