2-Methoxy-p-cresol 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Creosol, 1-Hydroxy-2-methoxy-4-methylbenzene, 2-Methoxy-p-cresol, 2-Methoxy-4-methylphenol, 4-Hydroxy-3-methoxytoluene, 4-Methylguaiacol, 2-Hydroxy-5-methylanisol
A13603-09
A13603-18
A13603-30
CAAAA13603-09EA
45.35
CAD
CAAAA13603-09
CAAAA13603-18
CAAAA13603-30
2-Methoxy-p-cresol 98+%
2-Methoxy-p-cresol
Purity: 98+%
CAS No.: 93-51-6
Molecular Formula: C8H10O2
Formula Weight: 138.17
Flash Point: 99°(210°F)
Refractive Index: 1.5370
Odor: Characteristic
Melting Point: 5-6°
Boiling Point: 220-222°
Beilstein Reference: 1862340
Merck Reference: 14,2571
MDL No.: MFCD00002378
Formula:
C₈H₁₀O₂ MW: 138.17 g/mol Boiling Pt: 220…222 °C Melting Pt: 5…6 °C Density: 1.092 Flash Pt: 99 °C (210 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002378 CAS Number: 93-51-6 EINECS: 202-252-9 Merck Index: 14,2571 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...