2,6-Difluorobenzenesulfonyl chloride 97%

Supplier: Thermo Scientific Chemicals
Danger

A13595-14 A13595-03 A13595-06
CAAAAA13595-14EA 1131.28 CAD
CAAAAA13595-14 CAAAA13595-03 CAAAA13595-06
2,6-Difluorobenzenesulfonyl chloride 97%
2,6-Difluorobenzenesulphonyl chloride

Purity: 97% CAS No.: 60230-36-6 Molecular Formula: C6H3ClF2O2S Formula Weight: 212.60 Flash Point: 110°(230°F) Refractive Index: 1.5260 Boiling Point: 127-130°/6.5mm UN No.: 3265 MDL No.: MFCD00085004

Formula: C₆H₃ClF₂O₂S
Boiling Pt: 127…130 °C/6.5 mm
Density: 1.568
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00085004
CAS Number: 60230-36-6
UN: 3265
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR