3-Nitrobenzaldehyde 99%

Supplier: Thermo Scientific Chemicals
Warning

A13594-0B A13594-30
CAAAA13594-0BEA 298.62 CAD
CAAAA13594-0B CAAAA13594-30
3-Nitrobenzaldehyde 99%
3-Nitrobenzaldehyde

Purity: 99% CAS No.: 99-61-6 Molecular Formula: C7H5NO3 Formula Weight: 151.12 Melting Point: 57-59° Boiling Point: 164°/23mm Merck Reference: 14,6587 MDL No.: MFCD00007249

Formula: C₇H₅NO₃
Boiling Pt: 164 °C/23 mm
Melting Pt: 57…59 °C
Density: 1.481 g/cm³ (0 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007249
CAS Number: 99-61-6
EINECS: 202-772-6
UN: 3077
ADR: 9,III
Merck Index: 14,6587

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR