4-(3-Phenylpropyl)pyridine 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A13590-0B
A13590-18
A13590-30
CAAAA13590-0BEA
432.92
CAD
CAAAA13590-0B
CAAAA13590-18
CAAAA13590-30
4-(3-Phenylpropyl)pyridine 98%
4-(3-Phenylpropyl)pyridine
Purity: 98%
CAS No.: 2057-49-0
Molecular Formula: C14H15N
Formula Weight: 197.28
Flash Point: 140°(284°F)
Refractive Index: 1.5630
Boiling Point: 320-322°
Beilstein Reference: 142242
MDL No.: MFCD00047458
Formula:
C₁₄H₁₅N Boiling Pt: 320…322 °C Density: 0,853 Flash Pt: 140 °C(284 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00047458 CAS Number: 2057-49-0 EINECS: 218-159-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...