Triethyl orthoformate 98%

Supplier: Thermo Scientific Chemicals
Warning

A13587-0D A13587-0F A13587-AP
CAAAA13587-0DEA 771.04 CAD
CAAAA13587-0D CAAAA13587-0F CAAAA13587-AP
Triethyl orthoformate 98%
Triethyl orthoformate

Purity: 98% CAS No.: 122-51-0 Molecular Formula: C7H16O3 Formula Weight: 148.20 Flash Point: 30°(86°F) Physical Form: Liquid Density: 0.891 Refractive Index: 1.3910 Sensitivity: Moisture Sensitive Appearance: Colorless Melting Point: -61° Boiling Point: 144-146° Beilstein Reference: 605384 Merck Reference: 14,6884 UN No.: 2524 MDL No.: MFCD00009230

Formula: C₇H₁₆O₃
MW: 148.2 g/mol
Boiling Pt: 144…146 °C
Melting Pt: –61 °C
Density: 0.891
Flash Pt: 30 °C (86 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009230
CAS Number: 122-51-0
EINECS: 204-550-4
UN: 2524
ADR: 3,III
Merck Index: 14,6884

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR