4-Aminophenol 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
p-Aminophenol, UrsolP Rodinal 4-Amino-1-hydroxybenzene p-Hyddroxyaniline 4-Aminophenol 4-Hydroxyaniline, 4-Hydroxyaniline
Purity: 98%
CAS No.: 123-30-8
Molecular Formula: C6H7NO
Formula Weight: 109.13
Flash Point: 195°(383°F)
Physical Form: Powder
Density: 1.29
Sensitivity: Air and Light Sensitive
Appearance: White, pale brown/purple
Odor: Phenol-like
Melting Point: 186-190°
Boiling Point: 284° dec.
Beilstein Reference: 385836
Merck Reference: 14,462
UN No.: 2512
MDL No.: MFCD00007869
Formula:
H₂NC₆H₄OH MW: 109.13 g/mol Boiling Pt: 284 °C (dec.) Melting Pt: 186…190 °C Density: 1.29 Flash Pt: 195 °C (383 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00007869 CAS Number: 123-30-8 EINECS: 204-616-2 UN: 2512 ADR: 6.1,III Merck Index: 14,462 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...