4-Hydroxybenzaldehyde 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A13580-14
A13580-22
A13580-36
CAAAAA13580-14EA
32.39
CAD
CAAAAA13580-14
CAAAA13580-22
CAAAA13580-36
4-Hydroxybenzaldehyde 98%
4-Hydroxybenzaldehyde
Purity: 98%
CAS No.: 123-08-0
Molecular Formula: C7H6O2
Formula Weight: 122.12
Flash Point: 174°(345°F)
Melting Point: 115-119°
Boiling Point: 191°/50mm
Beilstein Reference: 471352
Merck Reference: 14,4811
MDL No.: MFCD00006939
Formula:
C₇H₆O₂ Boiling Pt: 191 °C/50 mm Melting Pt: 115…119 °C Density: 1.129 Flash Pt: 174 °C(345 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00006939 CAS Number: 123-08-0 EINECS: 204-599-1 Merck Index: 14,4811 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...