Rhodamine B
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
9-(2-Carboxyphenyl)-3,6-bis(diethylamino)xanthylium chloride, [9-(2-Carboxyphenyl)-6-(diethylamino)xanthen-3-ylidene]-diethylazanium chloride
Suitable for fluorescent labelling
CAS No.: 81-88-9
Molecular Formula: C28H31ClN2O3
Formula Weight: 479.02
Physical Form: Crystals
Density: 1.31
Appearance: Purple powder/dark green
Melting Point: ca 210° dec.
Beilstein Reference: 4119648
Merck Reference: 14,8183
MDL No.: MFCD00011931
Suitable for fluorescent labelling
Formula:
C₂₈H₃₁ClN₂O₃ MW: 479.02 g/mol Melting Pt: ∼ 210 °C (dec.) Density: 1.31 Storage Temperature: Ambient |
MDL Number:
MFCD00011931 CAS Number: 81-88-9 EINECS: 201-383-9 Merck Index: 14,8183 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...