trans-2-Hexenoic acid 96%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A13571-09
A13571-18
CAAAA13571-09EA
53.4
CAD
CAAAA13571-09
CAAAA13571-18
trans-2-Hexenoic acid 96%
trans-2-Hexenoic acid
Purity: 96%
CAS No.: 13419-69-7
Molecular Formula: C6H10O2
Formula Weight: 114.14
Flash Point: 125°(257°F)
Refractive Index: 1.4385
Melting Point: 30-35°
Boiling Point: 217°
Beilstein Reference: 1720443
UN No.: 3261
MDL No.: MFCD00002705
Formula:
C₆H₁₀O₂ Boiling Pt: 217 °C Melting Pt: 30…35 °C Density: 0.965 Flash Pt: 125 °C (257 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002705 CAS Number: 13419-69-7 EINECS: 236-528-5 UN: 3261 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...