3',4'-Dichloroacetophenone 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A13568-14
A13568-22
CAAAA13568-14EA
87.06
CAD
CAAAA13568-14
CAAAA13568-22
3',4'-Dichloroacetophenone 98+%
3',4'-Dichloroacetophenone
Purity: 98+%
CAS No.: 2642-63-9
Molecular Formula: C8H6Cl2O
Formula Weight: 189.04
Melting Point: 72-76°
Boiling Point: 134-135mm/12mm
MDL No.: MFCD00000553
Formula:
C₈H₆Cl₂O Boiling Pt: 134…135 mm/12 mm Melting Pt: 72…76 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000553 CAS Number: 2642-63-9 EINECS: 220-146-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...