2-Iodobenzoyl chloride 98%

Supplier: Thermo Scientific Chemicals
Danger

A13558-09 A13558-18
CAAAAA13558-30EA 2050.63 CAD
CAAAAA13558-30 CAAAA13558-09 CAAAA13558-18
2-Iodobenzoyl chloride 98%
2-Iodobenzoyl chloride

Purity: 98% CAS No.: 609-67-6 Molecular Formula: C7H4ClIO Formula Weight: 266.47 Flash Point: 110°(230°F) Melting Point: 27-32° Boiling Point: 105-106°/1mm Beilstein Reference: 2042672 UN No.: 3261 MDL No.: MFCD00001040

Formula: C₇H₄ClIO
Boiling Pt: 105…106 °C/1 mm
Melting Pt: 27…32 °C
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001040
CAS Number: 609-67-6
EINECS: 210-196-1
UN: 3261
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR