Heptyl-4-hydroxybenzoate 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Heptylparaben, n-Heptyl 4-hydroxybenzoate, Heptyl parahydroxybenzoate, 4-Hydroxybenzoic acid heptyl ester

A13555-14 A13555-22
CAAAA13555-14EA 165.9 CAD
CAAAA13555-14 CAAAA13555-22
Heptyl-4-hydroxybenzoate 98%
Heptyl 4-hydroxybenzoate

Purity: 98% CAS No.: 1085-12-7 Molecular Formula: C14H20O3 Formula Weight: 236.31 Melting Point: 46-49° MDL No.: MFCD00016481

Formula: C₁₄H₂₀O₃
MW: 236.31 g/mol
Melting Pt: 46…49 °C
Storage Temperature: Ambient
MDL Number: MFCD00016481
CAS Number: 1085-12-7
EINECS: 214-115-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR