4-Chloro-3-nitrobenzaldehyde 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A13549-09
A13549-18
A13549-30
CAAAA13549-09EA
78.05
CAD
CAAAA13549-09
CAAAA13549-18
CAAAA13549-30
4-Chloro-3-nitrobenzaldehyde 97%
4-Chloro-3-nitrobenzaldehyde
Purity: 97%
CAS No.: 16588-34-4
Molecular Formula: C7H4ClNO3
Formula Weight: 185.57
Melting Point: 61-65°
Beilstein Reference: 778323
Formula:
C₇H₄ClNO₃ Melting Pt: 61…65 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007078 CAS Number: 16588-34-4 EINECS: 240-645-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...