3,4-Dimethoxyaniline 98+%

Supplier: Thermo Scientific Chemicals
Warning

A13548-09 A13548-18 A13548-30
CAAAA13548-09EA 72.05 CAD
CAAAA13548-09 CAAAA13548-18 CAAAA13548-30
3,4-Dimethoxyaniline 98+%
3,4-Dimethoxyaniline

Purity: 98+% CAS No.: 6315-89-5 Molecular Formula: C8H11NO2 Formula Weight: 153.18 Flash Point: 110°(230°F) Odor: Characteristic Boiling Point: 174-176°/22mm Beilstein Reference: 743399 UN No.: 2811 MDL No.: MFCD00008394

Formula: C₈H₁₁NO₂
MW: 153.18 g/mol
Boiling Pt: 174…176 °C/22 mm
Melting Pt: 86…89 °C
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008394
CAS Number: 6315-89-5
EINECS: 228-647-6
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR