2,4-Dihydroxybenzoic acid 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A13545-0E
A13545-22
A13545-36
CAAAAA13545-0EEA
462.94
CAD
CAAAAA13545-0E
CAAAA13545-22
CAAAA13545-36
2,4-Dihydroxybenzoic acid 97%
2,4-Dihydroxybenzoic acid
Purity: 97%
CAS No.: 89-86-1
Molecular Formula: C7H6O4
Formula Weight: 154.12
Melting Point: ca 226° dec.
Beilstein Reference: 1946213
Merck Reference: 14,8157
MDL No.: MFCD00002451
Formula:
C₇H₆O₄ Boiling Pt: 390 °C (1013 hPa) Melting Pt: ca 226 °C (dec.) Density: 1.54 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00002451 CAS Number: 89-86-1 EINECS: 201-946-9 Merck Index: 14,8157 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...