3-Hydroxybenzaldehyde 97%

Supplier: Thermo Scientific Chemicals
Warning

A13541-14 A13541-22 A13541-36
CAAAAA13541-14EA 38.39 CAD
CAAAAA13541-14 CAAAA13541-22 CAAAA13541-36
3-Hydroxybenzaldehyde 97%
3-Hydroxybenzaldehyde

Purity: 97% CAS No.: 100-83-4 Molecular Formula: C7H6O2 Formula Weight: 122.12 Melting Point: 102-105° Boiling Point: 191°/50mm MDL No.: MFCD00003368

Formula: C₇H₆O₂
MW: 122.12 g/mol
Boiling Pt: 191 °C/50 mm
Melting Pt: 102…105 °C
Density: 1,1299 g/cm³ (114 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003368
CAS Number: 100-83-4
EINECS: 202-892-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR