Cyclopropyl methyl ketone 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 1-Cyclopropylethanone

A13540-36 A13540-14 A13540-22
CAAAAA13540-36EA 680.98 CAD
CAAAAA13540-36 CAAAA13540-14 CAAAA13540-22
Cyclopropyl methyl ketone 99%
Cyclopropyl methyl ketone

Purity: 99% CAS No.: 765-43-5 Molecular Formula: C5H8O Formula Weight: 84.12 Flash Point: 16°(61°F) Refractive Index: 1.4240 Melting Point: -70° Boiling Point: 113-114° Beilstein Reference: 1633817 UN No.: 1992 MDL No.: MFCD00001297

Formula: C₅H₈O
MW: 84.12 g/mol
Boiling Pt: 114
Melting Pt: -68
Density: 0.898
Flash Pt: 16 °C (61 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001297
CAS Number: 765-43-5
EINECS: 212-146-4
UN: 2924
ADR: 3 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR