4'-Nitroacetanilide 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: N-(4-Nitrophenyl)acetamide

A13539-06 A13539-14
CAAAA13539-06EA 46.14 CAD
CAAAA13539-06 CAAAA13539-14
4'-Nitroacetanilide 98%
4'-Nitroacetanilide

Purity: 98% CAS No.: 104-04-1 Molecular Formula: C8H8N2O3 Formula Weight: 180.16 Melting Point: 215-217° Beilstein Reference: 2211962 Merck Reference: 14,6581 MDL No.: MFCD00007303

Formula: C₈H₈N₂O₃
MW: 180.16 g/mol
Boiling Pt: 409
Melting Pt: 215…217 °C
Storage Temperature: Ambient
MDL Number: MFCD00007303
CAS Number: 104-04-1
EINECS: 203-169-0
Merck Index: 14,6581

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR