trans-Cinnamic acid 99+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A13538-0E
A13538-22
A13538-36
CAAAA13538-0EEA
613.04
CAD
CAAAA13538-0E
CAAAA13538-22
CAAAA13538-36
trans-Cinnamic acid 99+%
trans-Cinnamic acid
Purity: 99+%
CAS No.: 140-10-3
Molecular Formula: C9H8O2
Formula Weight: 148.16
Flash Point: 166°(330°F)
Physical Form: Powder
Density: 1.250
Appearance: White to pale cream
Odor: Weak, characteristic
Melting Point: 133-136°
Boiling Point: 147°/4mm
Beilstein Reference: 1905952
Merck Reference: 14,2299
MDL No.: MFCD00004369
Formula:
C₉H₈O₂ Boiling Pt: 147 °C/4 mm Melting Pt: 133…136 °C Density: 1.250 Flash Pt: 166 °C(330 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004369 CAS Number: 140-10-3 EINECS: 205-398-1 Merck Index: 14,2299 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...