Benzyl bromide 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A13535-0B
A13535-18
A13535-30
CAAAA13535-0BEA
627.26
CAD
CAAAA13535-0B
CAAAA13535-18
CAAAA13535-30
Benzyl bromide 99%
Benzyl bromide
Purity: 99%
CAS No.: 100-39-0
Molecular Formula: C7H7Br
Formula Weight: 171.04
Flash Point: 87°(189°F)
Physical Form: Liquid
Density: 1.438
Refractive Index: 1.5750
Sensitivity: Moisture Sensitive
Odor: Pungent, makes eyes water
Melting Point: -4°
Boiling Point: 198-199°
Beilstein Reference: 385801
Merck Reference: 14,1128
UN No.: 1737
MDL No.: MFCD00000172
Formula:
(C₆H₅)CH₂Br Boiling Pt: 198…199 °C Melting Pt: –4 °C Density: 1.438 Flash Pt: 87 °C (189 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000172 CAS Number: 100-39-0 EINECS: 202-847-3 UN: 1737 ADR: 6.1,II Merck Index: 14,1128 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...