2-Naphthylacetic acid 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Naphthaleneacetic acid, 2-(Naphthalen-2-yl)acetic acid

A13528-06 A13528-09 A13528-18
CAAAAA13528-06EA 94.8 CAD
CAAAAA13528-06 CAAAA13528-09 CAAAA13528-18
2-Naphthylacetic acid 99%
2-Naphthylacetic acid

Purity: 99% CAS No.: 581-96-4 Molecular Formula: C12H10O2 Formula Weight: 186.21 Melting Point: 141-143° Beilstein Reference: 973396

Formula: C₁₂H₁₀O₂
MW: 186.21 g/mol
Boiling Pt: 352 °C (1013 hPa)
Melting Pt: 141…143 °C
Density: 1.29 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004126
CAS Number: 581-96-4
EINECS: 209-475-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR