3,5-Dichlorobenzyl alcohol 98%

Supplier: Thermo Scientific Chemicals
Warning

A13527-06 A13527-14
CAAAA13527-06EA 74.73 CAD
CAAAA13527-06 CAAAA13527-14
3,5-Dichlorobenzyl alcohol 98%
3,5-Dichlorobenzyl alcohol

Purity: 98% CAS No.: 60211-57-6 Molecular Formula: C7H6Cl2O Formula Weight: 177.03 Melting Point: 76-80° Beilstein Reference: 1936389

Formula: C₇H₆Cl₂O
Boiling Pt: 264 °C (1013 hPa)
Melting Pt: 76…80 °C
Storage Temperature: Ambient
MDL Number: MFCD00004634
CAS Number: 60211-57-6
EINECS: 262-109-1
UN: 2250
ADR: 6.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR