2-Aminobenzothiazole 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
Benzothiazol-2-ylamine, Benzo[d]thiazol-2-amine, Cerium(III)-Ionophore II
A13521-22
A13521-36
CAAAA13521-22EA
68.1
CAD
CAAAA13521-22
CAAAA13521-36
2-Aminobenzothiazole 98%
2-Aminobenzothiazole
Purity: 98%
CAS No.: 136-95-8
Molecular Formula: C7H6N2S
Formula Weight: 150.20
Melting Point: 126-129°
Beilstein Reference: 116315
Merck Reference: 14,424
MDL No.: MFCD00005785
Formula:
C₇H₆N₂S MW: 150.2 g/mol Boiling Pt: 190…195 °C (0.05 mmHg) Melting Pt: 126…129 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005785 CAS Number: 136-95-8 EINECS: 205-268-4 Merck Index: 14,424 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...