L(+)-Aspartic acid 98+%

Supplier: Thermo Scientific Chemicals

A13520-0B A13520-0I
CAAAAA13520-22EA 38.14 CAD
CAAAAA13520-22 CAAAA13520-0B CAAAA13520-0I
L(+)-Aspartic acid 98+%
L(+)-Aspartic acid

Purity: 98+% CAS No.: 56-84-8 Molecular Formula: C4H7NO4 Formula Weight: 133.10 Odor: Odorless Melting Point: 300° dec. Beilstein Reference: 1723530 Merck Reference: 14,840 Optical Rotation: +25° (c=2 in 5N HCl) MDL No.: MFCD00002616

Formula: C₄H₇NO₄
Boiling Pt: 311 °C (1013 hPa)
Melting Pt: >300 °C (dec.)
Density: 1.66
Storage Temperature: Ambient
MDL Number: MFCD00002616
CAS Number: 56-84-8
EINECS: 200-291-6
Merck Index: 14,840

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR