o-Anisic acid 98+%

Supplier: Thermo Scientific Chemicals
Warning

A13513-22 A13513-36
CAAAA13513-22EA 78.05 CAD
CAAAA13513-22 CAAAA13513-36
o-Anisic acid 98+%
o-Anisic acid

Purity: 98+% CAS No.: 579-75-9 Molecular Formula: C8H8O3 Formula Weight: 152.15 Odor: Odorless Melting Point: 99-103° Beilstein Reference: 509929 MDL No.: MFCD00002431

Formula: C₈H₈O₃
Boiling Pt: 280 °C (1013 hPa)
Melting Pt: 99…103 °C
Density: 1.38 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002431
CAS Number: 579-75-9
EINECS: 209-447-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR