3-Chlorobenzoyl chloride 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A13509-14
A13509-22
CAAAAA13509-36EA
865.87
CAD
CAAAAA13509-36
CAAAA13509-14
CAAAA13509-22
3-Chlorobenzoyl chloride 97%
3-Chlorobenzoyl chloride
Purity: 97%
CAS No.: 618-46-2
Molecular Formula: C7H4Cl2O
Formula Weight: 175.01
Flash Point: 117°(242°F)
Refractive Index: 1.5690
Odor: Acidic
Boiling Point: 224-225°
Beilstein Reference: 386256
UN No.: 3265
MDL No.: MFCD00000671
Formula:
C₇H₄Cl₂O Boiling Pt: 224…225 °C Melting Pt: 44 °C Density: 1.368 Flash Pt: 117 °C (242 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000671 CAS Number: 618-46-2 EINECS: 210-552-6 UN: 3265 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...