1-Cetylpyridinium chloride monohydrate 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Hexadecylpyridinium chloride monohydrate, 1-Hexadecylpyridinium chloride monohydrate, Cetylpyridinium Chloride, Monohydrate, Cetylpyridinium chloride monohydrate
MDL: MFCD00149977
Beilstein: 3578606
Soluble in water, alcohol, chloroform.
Purity: 98%
CAS No.: 6004-24-6
Molecular Formula: C21H38ClN.H2O
Formula Weight: 358.02 (339.99anhy)
Melting Point: 83-86°
Solubility: Soluble in water, alcohol, chloroform
Beilstein Reference: 3578606
Merck Reference: 14,2032
UN No.: 2811
MDL No.: MFCD00149977
Formula:
C₂₁H₃₈ClN·1H₂O MW: 358.01 g/mol Melting Pt: 83…86 °C Storage Temperature: Ambient |
MDL Number:
MFCD00149977 CAS Number: 6004-24-6 EINECS: 204-593-9 UN: 2811 ADR: 6.1,III Merck Index: 14,2032 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...