2-Chlor-4-fluorothiophenol 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2-Chlor-4-fluorobenzene-1-thiol, 2-Chloro-4-fluorobenzenethiol, 1-Chloro-3-fluoro-6-mercaptobenzene, 2-Chlor-4-fluorobenzenethiol
A13498-03
A13498-06
CAAAA13498-03EA
107.28
CAD
CAAAA13498-03
CAAAA13498-06
2-Chlor-4-fluorothiophenol 97%
2-Chlor-4-fluorothiophenol
Purity: 97%
CAS No.: 175277-99-3
Molecular Formula: C6H4ClFS
Formula Weight: 162.61
Refractive Index: 1.5670
Boiling Point: 86-88°/15mm
MDL No.: MFCD00052936
Formula:
C₆H₄ClFS MW: 162.61 g/mol Boiling Pt: 86…88 °C/15 mm Storage Temperature: Ambient |
MDL Number:
MFCD00052936 CAS Number: 175277-99-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...