2,3-Dihydroxybenzaldehyde 97%

Supplier: Thermo Scientific Chemicals
Warning

A13490-22 A13490-06 A13490-14
CAAAAA13490-22EA 995.4 CAD
CAAAAA13490-22 CAAAA13490-06 CAAAA13490-14
2,3-Dihydroxybenzaldehyde 97%
2,3-Dihydroxybenzaldehyde

Purity: 97% CAS No.: 24677-78-9 Molecular Formula: C7H6O3 Formula Weight: 138.12 Melting Point: 106-109° Beilstein Reference: 2041781

Formula: C₇H₆O₃
Boiling Pt: 235 °C (1013 hPa)
Melting Pt: 106…109 °C
Storage Temperature: Ambient
MDL Number: MFCD00003324
CAS Number: 24677-78-9
EINECS: 246-398-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR