3-Chloro-1-iodopropane 98% stabilized

Supplier: Thermo Scientific Chemicals
Warning


Stabilization: Stabilised with copper

A13489-14 A13489-22 A13489-36
CAAAA13489-14EA 97.49 CAD
CAAAA13489-14 CAAAA13489-22 CAAAA13489-36
3-Chloro-1-iodopropane 98% stabilized
3-Chloro-1-iodopropane

Purity: 98% CAS No.: 6940-76-7 Molecular Formula: ClCH2CH2CH2I Formula Weight: 204.44 Flash Point: 110°(230°F) Refractive Index: 1.5463 Boiling Point: 170-172° Beilstein Reference: 1731115 MDL No.: MFCD00001096

Formula: C₃H₆ClI
Boiling Pt: 170…172 °C
Density: 1.904
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001096
CAS Number: 6940-76-7
EINECS: 230-088-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR