2-Chlorobenzaldehyde 97%

Supplier: Thermo Scientific Chemicals
Danger

A13488-09 A13488-0B A13488-30
CAAAAA13488-09EA 37.45 CAD
CAAAAA13488-09 CAAAA13488-0B CAAAA13488-30
2-Chlorobenzaldehyde 97%
2-Chlorobenzaldehyde

Purity: 97% CAS No.: 89-98-5 Molecular Formula: C7H5ClO Formula Weight: 140.57 Flash Point: 87°(188°F) Refractive Index: 1.5675 Odor: Pungent pH: 2.9 Melting Point: 10-12° Boiling Point: 212° Beilstein Reference: 385877 UN No.: 3265 MDL No.: MFCD00003304

Formula: C₇H₅ClO
Boiling Pt: 212 °C
Melting Pt: 10…12 °C
Density: 1.249
Flash Pt: 87 °C(188 °F)
Storage Temperature: Ambient
MDL Number: MFCD00003304
CAS Number: 89-98-5
EINECS: 201-956-3
UN: 3265
ADR: "8",III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR