Bicyclohexyl 98%

Supplier: Thermo Scientific Chemicals
Warning

A13481-09 A13481-18
CAAAA13481-09EA 83.27 CAD
CAAAA13481-09 CAAAA13481-18
Bicyclohexyl 98%
Bicyclohexyl

Purity: 98% CAS No.: 92-51-3 Molecular Formula: C12H22 Formula Weight: 166.31 Flash Point: 92°(198°F) Refractive Index: 1.478 Melting Point: 3-4° Boiling Point: 226-228° Beilstein Reference: 1848266 MDL No.: MFCD00003815

Formula: C₁₂H₂₂
MW: 166.31 g/mol
Boiling Pt: 226…228 °C
Melting Pt: 3…4 °C
Density: 0.864
Flash Pt: 92 °C(198 °F)
Storage Temperature: Ambient
MDL Number: MFCD00003815
CAS Number: 92-51-3
EINECS: 202-161-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR