Iron(II) oxalate dihydrate 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A13479-0B
A13479-30
CAAAA13479-0BEA
271.76
CAD
CAAAA13479-0B
CAAAA13479-30
Iron(II) oxalate dihydrate 99%
Iron (II) oxalate dihydrate
Purity: 99%
CAS No.: 6047-25-2
Molecular Formula: FeC2O4.2H2O
Formula Weight: 179.90 (143.87anhy)
Physical Form: Crystals
Density: 2.28
Appearance: Yellow
Melting Point: 190° dec.
MDL No.: MFCD00150040
Formula:
Fe(C₂O₄)·2H₂O Melting Pt: 190 °C (dec.) Density: 2.28 Storage Temperature: Ambient |
MDL Number:
MFCD00150040 CAS Number: 6047-25-2 EINECS: 208-217-4 UN: 3288 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...