5-Nitro-1H-indazole 98+%
Supplier: Thermo Scientific Chemicals
A13478-14
A13478-22
A13478-36
CAAAA13478-14EA
124.5
CAD
CAAAA13478-14
CAAAA13478-22
CAAAA13478-36
5-Nitro-1H-indazole 98+%
5-Nitro-1H-indazole
Purity: 98+%
CAS No.: 5401-94-5
Molecular Formula: C7H5N3O2
Formula Weight: 163.14
Odor: Odorless
Melting Point: 208-211°
Beilstein Reference: 7936
MDL No.: MFCD00005693
Formula:
C₇H₅N₃O₂ Melting Pt: 208…211 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005693 CAS Number: 5401-94-5 EINECS: 226-451-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...